Learn More
Invitrogen™ Destrin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529933
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, A549, H1299, HCT116, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (95%), Pig (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| Destrin | |
| Polyclonal | |
| Unconjugated | |
| DSTN | |
| 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11034, 56431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P60981, Q9R0P5 | |
| DSTN | |
| Recombinant protein encompassing a sequence within the center region of human Destrin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.