Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Destrin Recombinant Rabbit Monoclonal Antibody (4F9Q1)
GREENER_CHOICE

Catalog No. PIMA537990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537990 Supplier Invitrogen™ Supplier No. MA537990
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: A-549, MCF7, NIH/3T3, C6, Mouse lung, Mouse spleen, Rat lung, Rat spleen Immunogen sequence: MASGVQVADE VCRIFYDMKV RKCSTPEEIK KRKKAVIFCL SADKKCIIVE EGKEILVGDV GVTITDPFKH FVGMLPEKDC RYALYDASFE TKESRKEELM.

The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Destrin
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4F9Q1
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DSTN
Gene Accession No. P60981, Q7M0E3, Q9R0P5
Gene Alias 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p
Gene Symbols DSTN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Destrin (P60981).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11034, 502674, 56431
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.