Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Desulfotalea psychrophila DNA ligase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117669
Description
This antibody has been tested in direct-ELISA.
DNA ligase that catalyzes the formation of phosphodiester linkages between 5'-phosphoryl and 3'-hydroxyl groups in double-stranded DNA using NAD as a coenzyme and as the energy source for the reaction. It is essential for DNA replication and repair of damaged DNA.
Specifications
| Desulfotalea psychrophila DNA ligase | |
| Polyclonal | |
| Unconjugated | |
| ligA | |
| ligA | |
| Recombinant Desulfotalea psychrophila DNA ligase protein (340-679aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 4.578 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P15042 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 57729416 | |
| -20°C or -80°C if preferred | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction