Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DET1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546710 Supplier Invitrogen™ Supplier No. PA546710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DCRCVIVGSA AYLPDEPHPP FFEVYRNSES VTPNPRSPLE DYSLHIIDLH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

DET1 is a component of the E3 ubiquitin ligase DCX DET1-COP1 complex, which is required for ubiquitination and subsequent degradation of target proteins. The complex is involved in JUN ubiquitination and degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DET1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DET1
Gene Accession No. Q7L5Y6
Gene Alias 2610034H20Rik; de-etiolated 1; de-etiolated homolog 1; de-etiolated homolog 1 (Arabidopsis); de-etiolated-1 homolog; DET1; DET1 homolog; DET1 partner of COP1 E3 ubiquitin ligase; DET1, COP1 ubiquitin ligase partner; RGD1309793; wu:fc18d06; zgc:66486
Gene Symbols DET1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DET1 (aa 150-199).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.