Learn More
Invitrogen™ DET1 Polyclonal Antibody

Description
Positive Samples: BT-474, Mouse thymus Immunogen sequence: HVSTIKPRRI QNQNVIHRLE RRRISSGKAG THWHQVRVFH QNVFPNFTVV NVEKPPCFLR KFSPDGRYFI AFSSDQTSLE IYEYQGCQAA EDLLQGYEGE ILSNGNDQRS VNIRGRLFER FFVLLHITNV AANGEHLNRE CSLFTDDCRC VIVGSAAYLP DEPHPPFFEV YRNSESVTPN PRSPLEDYSL HIIDLHTGRL CDTRTFKCDK VVLSHNQGLY LYKNIL.
Specifications
Specifications
| Antigen | DET1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DET1 |
| Gene Accession No. | Q7L5Y6, Q9D0A0 |
| Gene Alias | 2610034H20Rik; de-etiolated 1; de-etiolated homolog 1; de-etiolated homolog 1 (Arabidopsis); de-etiolated-1 homolog; DET1; DET1 homolog; DET1 partner of COP1 E3 ubiquitin ligase; DET1, COP1 ubiquitin ligase partner; RGD1309793; wu:fc18d06; zgc:66486 |
| Gene Symbols | DET1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.