Description
- Do not use past expiration date.
- Supplied ready for use.
- If the Detergent Reagent has been stored at 4°C, warm the bottle for 5 minutes at 37°C then invert gently while mixing to avoid frothing.
- Contains SDS which is an irritant. In case of contact, immediately flush eyes or skin with copious amounts of water. If swallowed, wash out mouth with water provided person is conscious.
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature |
| Product Line | R&D Systems |
| Quantity | 250 mL |