Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Columns, 1 mL PROMO

Catalog No. PI20344PM
Click to view available options
:
Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Columns

Bind and extract endotoxins (lipopolysaccharides or LPS) from antibody or protein samples using bulk resin or columns of immobilized Polymixin B ligand.

Detoxi-Gel Resin is a beaded agarose support onto which polymixin B has been immobilized. This ligand binds to the lipid A portion of lipopolysaccharides (LPS), which are the dominant form of endotoxins that affect biological samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be added to cells for various treatment experiments. Detoxi-Gel Resin provides a quick method for removal of endotoxins from samples. It is available in convenient pre-packed columns and two sizes of slurry for use in small-scale or large-scale processes.

Highlights:

  • Affinity-based – uses immobilized polymixin B to remove endotoxins by binding to their lipid A domains

  • High capacity – 1 mL of Detoxi-Gel Resin removes at least 9995 endotoxin units from a 5 mL sample containing 10,000 EU of lipopolysaccharide (LPS)

  • Reusable – no loss in binding capacity even after 10 regenerations; requires activation and regeneration with 1% sodium deoxycholate (Part No. 89904)

  • Testing required – protein recovery is dependent upon protein type and concentration; empirical testing is required to validate the method for particular protein samples

  • Versatile – remove endotoxin from protein solutions, cell culture media and pharmacological components

Requires:

Sodium deoxycholate (Part No. 89904) for resin activation and regeneration.

Specifications

Capacity (Metric) 1 mL
Content And Storage Store at 4°C.
Form Liquid
Product Line Detoxi-Gel™
Quantity 5 Columns
Target Endotoxin Removal
Type Endotoxin Removing Column
Stationary Phase Polymixin B
Format Spin Column
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.