Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Devardas alloy, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 g
1 kg
Description
Food and Beverages, Forensics and Toxicology

Specifications
Specifications
Melting Point | 550°C |
Quantity | 1 kg |
CAS | 8049-11-4 |
Form | Powder |
Linear Formula | AlCuZn |
Grade | puriss. p.a. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction