Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DeviceLink Connect Hardware

The DeviceLink Connect is a remote monitoring and asset management solution for laboratory equipment enabling knowledge based decisions by monitoring critical equipment parameters while providing data insights and real-time system notifications.

$4.71 - $785.50

Specifications

For Use With (Application) Wireless Monitoring, Asset Management Solution
View More Specs

Products 6
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
DLCPTBRKT1
View Documents
Thermo Scientific™
DLCPTBRKT001
LN2 Temperature sensor mounting bracket LN2 Cryo chambers
Each for $166.80
Only null left
Add to Cart
 
DLCPTETHE3
View Documents
Thermo Scientific™
DLCPTETHE003
Ethernet (LAN) cable DeviceLink Connect mote
Each for $20.45
Only null left
Add to Cart
 
DLCPTMONT1
View Documents
Thermo Scientific™
DLCPTMONT001
Cable mounting hardware (spare parts) DeviceLink Connect system
Each for $4.71
Only null left
Add to Cart
 
DLCPTMOTE1
View Documents
Thermo Scientific™
DLCPTMOTE001
DeviceLink Connect mote (only) DeviceLink Connect system
Each for $785.50
Only null left
Add to Cart
 
DLCPTPWRC1
View Documents
Thermo Scientific™
DLCPTPWRC001
DeviceLink Connect mote
Each for $100.30
Only null left
Add to Cart
 
DLCSWCOMM1
View Documents
Thermo Scientific™
DLCSWCOMM001
USB Commissioning (setup) dongle DeviceLink Connect mote
Each for $20.35
Only null left
Add to Cart
 
Description

Description

Thermo Scientific™ DeviceLink Connect is the newest addition to Thermo Scientific’s DeviceLink portfolio that measures critical reliability parameters and uses advanced analytics to provide real-time monitoring and reporting for lab instruments. The system supports independent sensors to monitor your instrument, but it can also be connected via serial port directly to the instrument harnessing its on-board telemetry data, alarms, and events. All information is transmitted to the Thermo Connect cloud where you have 24/7 access to the information via web and mobile based platforms.

The DeviceLink Connect is a remote monitoring system that uses direct serial/USB based instrument connections and/or independent sensors to gather useful information about the operation of the equipment. Sending this data back to Thermo Connect cloud allows for 24/7 monitoring from anywhere in the world.

How it works:

  1. Connect your DeviceLink Connect system
    • Supply power via the provided AC power cable
    • Connect your instrument via serial/USB cable to the DeviceLink Connect mote
    • Install (optional) independent sensors
  2. Configure your setup via the web-based Commissioner Application
  3. Establish a Thermo Connect cloud account
  4. Download the free mobile InstrumentConnect mobile application
  5. Monitor your equipment 24/7 from anywhere in the world

Network:

  • Wi-Fi: WPA Enterprise (802.1x) and WPA2-PSK protocols
  • Ethernet (LAN) connections also supported
  • Up to 7 days internal data storage during a network outage

Power and Battery:

  • Power-over-Ethernet (PoE) provides main power to the device.
  • PoE wall adapter provide or a PoE 802.3af standard connection can be used
  • Contains a rechargeable Li-ion backup battery providing up to 3 hrs. of backup power during continuous operation (2600 mAH, 3.6V)

Advanced InSight Analytics:

The DeviceLink Connect system is also compatible with Thermo Scientific™ InSight™ analytics, powered by KLATU™ which are designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:

  • Cabinet temperature
  • Electrical use and energy consumption
  • Door openings
  • Ambient temperature

When you purchase an InSight monitoring license and subscription (sold separately) your DeviceLink Connect data run through a number of algorithms utilizing the InSight software. With InSight analytics, you can determine and compare the assets' performance against industry performance standards adding that next level of asset management.

Installation and Services:

  • Customer (self) installation
  • Install services are sold separately if desired
  • Calibration sensors also available.

Available only in North America.

Specifications

Specifications

Wireless Monitoring, Asset Management Solution
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.