Learn More
Thermo Scientific™ DeviceLink Connect Remote Monitoring System (Kits) PROMO
The DeviceLink Connect is a remote monitoring and asset management solution for laboratory equipment, enabling knowledge-based decisions by monitoring critical equipment parameters while providing data insights and real-time system notifications.
Supplier: Thermo Scientific™ DLCKTULTS101P
Description
The DeviceLink Connect is a remote monitoring system that uses direct serial/USB based instrument connections and/or independent sensors to gather useful information about the operation of the equipment. Sending this data back to Thermo Connect cloud allows for 24/7 monitoring from anywhere in the world. Alarms by email, in-application, or mobile phone.
How it works:
- Connect your DeviceLink Connect system
- Supply power via the provided AC power cable
- Connect your instrument via serial/USB cable to the DeviceLink Connect mote
- Install (optional) independent sensors
- Configure your setup via the web-based Commissioner Application
- Establish a Thermo Connect cloud account
- Download the free mobile InstrumentConnect mobile application
- Monitor your equipment 24/7 from anywhere in the world
Network:
- Wi-Fi: WPA Enterprise (802.1x) and WPA2-PSK protocols
- Ethernet (LAN) connections also supported
- Up to 7 days internal data storage during a network outage
Power and Battery:
- Power-over-Ethernet (PoE) provides main power to the device
- PoE wall adapter provide or a PoE 802.3af standard connection can be used
- Contains a rechargeable Lithium ion backup battery providing up to 3 hr of backup power during continuous operation (2600 mAH, 3.6 V)
Advanced InSight Analytics:
The DeviceLink Connect system is also compatible with Thermo Scientific™ InSight™ analytics, powered by KLATU™ which are designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
- Cabinet temperature
- Electrical use and energy consumption
- Door openings
- Ambient temperature
When you purchase an InSight™ monitoring license and subscription (sold separately) your DeviceLink Connect data run through a number of algorithms utilizing the InSight™ software. With InSight™ analytics, you can determine and compare the assets' performance against industry performance standards adding that next level of asset management.
Installation and Services:
- Customer (self) installation
- Install services are sold separately if desired
- Calibration sensors also available.
Available only in North America.
Specifications
Lithium-ion | |
DeviceLink Connect Mote, PoE Power Supply, USB Commissioning Dongle, Serial to USB ULT Connection Cable, Cable Mounting Ties, First Year DeviceLink Connect Subscription | |
Serial to Equipment, Temperature, Humidity, Current, CO2, Door Switch | |
Thermo Scientific™ ULT Freezers, All Lab Equipment |
3 hr | |
Ethernet (LAN) or Wi-Fi Connections | |
Wireless Monitoring, Asset Management Solution |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.