Learn More
Epredia™ Dewax and HIER Buffer L,M,H
Catalog No. p-4670463 Shop All Epredia Products
Click to view available options
Solution Type:
Buffer - High
Buffer - Low
Buffer - Medium
Catalog No. TA999DHBH
Supplier: Epredia™ TA999DHBH
Only null left
Add to Cart
Add to Cart
Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.
Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.
Specifications
Product Type Dewax and HIER Buffer
Quantity 10 x 100 mL
Solution Type Buffer - High
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.