Learn More
Synthware™ Dewer Style Vacuum Trap
GREENER_CHOICESDP
Dry ice, dewar syle trap condenser designed for large volume solvent removal
$715.80 - $804.00
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Quantity 1 Ea.
View More Specs
Products 6
Catalog Number Mfr. No. Type Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Type Capacity (Metric) Price Quantity & Availability  
31-501-842
GREENER_CHOICESDP
View Documents
Synthware™
V210250G
Glass Stopcock 250 mL
Each for $715.80
Only null left
Add to Cart
31-501-843
GREENER_CHOICESDP
View Documents
Synthware™
V210250
PTFE Stopcock 250 mL
Each for $731.40
Only null left
Add to Cart
31-501-844
GREENER_CHOICESDP
View Documents
Synthware™
V210500G
Vacuum Trap - Dewar 500 mL
Each for $719.40
Only null left
Add to Cart
31-501-845
GREENER_CHOICESDP
View Documents
Synthware™
V210500
PTFE Stopcock 500 mL
Each for $784.20
Only null left
Add to Cart
31-501-846
GREENER_CHOICESDP
View Documents
Synthware™
V211000G
Glass Stopcock 1000 mL
Each for $756.00
Only null left
Add to Cart
31-501-847
GREENER_CHOICESDP
View Documents
Synthware™
V211000
PTFE Stopcock 1000 mL
Each for $804.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Organic Synthesis
Borosilicate Glass
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.