Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone 21-Phosphate Disodium Salt, MP Biomedicals™

Catalog No. mp215756594 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g

Catalog No. MP215756594

Supplier: MP Biomedicals, Inc 0215756594

Only null left
Add to Cart
Add to Cart

Induces apoptosis and inhibits sodium phosphate symporter. Dexamethasone 21-Phosphate Disodium Salt, MP Biomedicals is a systemic corticosteroid that is converted to dexamethasone in vivo. It is an anti-inflammatory glucocorticoid, a pro-drug that stimula

Dexamethasone Sodium Phosphate is used in the treatment of emetic effects of cancer therapy and epicondylitis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2392-39-4
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
InChI Key PLCQGRYPOISRTQ-FCJDYXGNSA-L
Synonym dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron
PubChem CID 16961
ChEBI CHEBI:4462
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water (200mg/4mL, clear, faint yellow solution).
Formula Weight 516.4
Percent Purity >97%
Recommended Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.