Learn More
Dexamethasone, 98%
Description
Anti-inflammatory glucocorticoidDexamethasone is used to treat inflammatory and autoimmune conditions such as rheumatoid arthritis and bronchospasm. It is useful to study apoptosis, cell signaling pathways and gene expression. It is associated with marbofloxacin and clotrimazole and finds application in veterinary medicine to treat difficult ear infections in dogs. It is also used to treat horses with swelling of of distal limbs and general bruising in combination with trichlormethiazide. It is also an anti-inflammatory glucocorticoid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anti-inflammatory glucocorticoidDexamethasone is used to treat inflammatory and autoimmune conditions such as rheumatoid arthritis and bronchospasm. It is useful to study apoptosis, cell signaling pathways and gene expression. It is associated with marbofloxacin and clotrimazole and finds application in veterinary medicine to treat difficult ear infections in dogs. It is also used to treat horses with swelling of of distal limbs and general bruising in combination with trichlormethiazide. It is also an anti-inflammatory glucocorticoid.
Solubility
Soluble in ethanol and dimethyl sulfoxide. Insoluble in water.
Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽260°C (decomposition) |
| Assay Percent Range | 0.98 |
| Quantity | 1 g |
| Beilstein | 2066651 |
| Sensitivity | Light sensitive |
| Merck Index | 142943 |
| Solubility Information | Soluble in ethanol and dimethyl sulfoxide. Insoluble in water. |
| Formula Weight | 392.47 |
| Percent Purity | 98% |
| Chemical Name or Material | Dexamethasone |
Safety and Handling
| Hazard Category | Carcinogenicity (category 2) |
| Hazard Statement | Suspected of causing cancer |
| EINECSNumber | 200-003-9 |
| RTECSNumber | TU3980000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.