Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dexamethasone Hemisuccinate Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533126
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533126 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533126 Supplier Invitrogen™ Supplier No. PA533126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dexamethasone Hemisuccinate Polyclonal Antibody for ELISA

Dexamethasone hemisuccinate is a synthetic steroid hormone often used as an immunosupressant or anti-inflammatory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dexamethasone Hemisuccinate
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias C26H33FO8; Deltasol
Host Species Rabbit
Immunogen Dexamethasone conjugated to Bovine Serum Albumin.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.