Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Dexamethasone
GREENER_CHOICESDP

Catalog No. 1126100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-261-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-261-00 Supplier R&D Systems™ Supplier No. 1126/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Anti-inflammatory glucocorticoid

Glucocorticoid; anti-inflammatory. Reduces levels of activated NF-κB in immature dendritic cells (DCs) and inhibits differentiation into mature DCs. Induces differentiation of human mesenchymal stem cells (MSCs). Also used in the generation of liver organoids. Induces autophagy in acute lymphoblastic leukemia (ALL) cell lines.

R&D Systems products are for biomedical research use only. They are not intended for human or veterinary use.

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 392.47
Purity >98%
Molecular Weight (g/mol) 392.47
Chemical Name or Material Dexamethasone
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.