Learn More
Description
Biological Activity: Indirectly agonizes serotonin receptors via inhibition of 5-HT re-uptake and stimulation of 5-HT release. Anorectic agent; decreases growth hormone, insulin, leptin, fat mass, lean mass and increases ghrelin in diet-switched diet-induced obese mice.
Specifications
Specifications
| CAS | 3239-45-0 |
| Molecular Formula | C12H16F3N.HCl |
| Solubility | Soluble to 100 mM in water and to 100 mM in DMSO |
| Content And Storage | Store at Room Temperature |
| Quantity | 50 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
