Learn More
Invitrogen™ DEXI Polyclonal Antibody

Description
Positive Samples: A-549, BxPC-3, HT-29 Immunogen sequence: MLGARVAAHL DALGPLVPYV PPPLLPSMFY VGLFFVNVLI LYYAFLMEYI VLNVGLVFLP EDMDQALVDL GVLSDPGSGL YDADSELDVF DAYLE.
Specifications
Specifications
| Antigen | DEXI |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Dexi |
| Gene Accession No. | O95424 |
| Gene Alias | 1810029J14Rik; AI836170; AW413143; D16Bwg0586e; Dexamethasone-induced protein; dexamethasone-induced transcript; Dexi; Dexi homolog; Myle; Protein MYLE; RGD1564938 |
| Gene Symbols | Dexi |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.