Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DEXRAS1, Mouse, Unlabeled, Clone: 4B11.1,

Catalog No. MABN313MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN313MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN313MI Supplier MilliporeSigma™ Supplier No. MABN313
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dexamethasone-induced Ras-related protein 1 (UniProt Q9Y272; also known as Activator of G-protein signaling 1) is encoded by the RASD1 (also known as AGS1, DEXRAS1) gene (Gene ID 51655) in human. Rasd1/Dexras belongs to the monomeric Ras superfamily of GTPases that are commonly referred to as small G-proteins. Rasd1/Dexras1 is a small G protein that functions as an important signal transducer in regulating circadian rhythms, GH secretion, and cell growth. Rasd1/Dexras is rapidly up-regulated by the synthetic glucocorticoid dexamethasone as a result of GR-STAT5b complex-mediated transcription. Rasd1/Dexras1 interacts with neuronal nitric oxide synthase (nNOS or bNOS) via the scaffolding protein CAPON, with nitric oxide serving as a guanine nucleotide exchange factor for Rasd1/Dexras1. Rasd1/Dexras1 is induced by glucocorticoids during adipogenic differentiation. Exogenous Rasd1/Dexras1 expression rescues impaired adipogenesis of Dexras1-deficient mouse embryonic fibroblasts (MEFs). Dexras1 knockout mice display impaired adiposity and are resistant to diet-induced weight gain. Human, moue, and rat Rasd1/Dexras1 protein is initially produced with a 3-amino acid propeptide sequence at the C-terminal end, which was posttranslationally removed to yield the mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEXRAS1
Applications Western Blot
Classification Monoclonal
Clone 4B11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RASD1, AGS1, DEXRAS1
Gene Accession No. Q9Y272
Host Species Mouse
Immunogen GST-tagged recombinant human DEXRAS1 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_057168
Test Specificity Clone 4B11.1 detects an epitope conserved among human, mouse, and rat RASD1 protein, but missing in human RASD1 spliced isoform 2 reported by UniProt (Q9Y272).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.