Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran sulfate sodium salt, approx. M.W. 500,000

Catalog No. p-4229023
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name Dextran sulfate sodium salt
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 5 g
Merck Index 15,2953
Solubility Information colorless to light yellow
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Chemical Name or Material Dextran sulfate sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.