Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran Sulfate Sodium Salt, MW ca 40,000

Catalog No. p-7042742
Click to view available options
Quantity:
25 g
100 g

Dextran sulfate sodium salt (benzocaine or sodium dextran sulfate, CAS # 94-09-7) is a glucose polymer of bacterial origin containing 17-20% sulfur. | CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It is widely used for laboratory applications including use in preparing a precipitating reagent for the determination of HDL Cholesterol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Dextran sulfate sodium salt is the ethyl ester of p-aminobenzoic acid
  • Dextran sulfate inhibits voltage-dependent sodium channels, preventing the transduction of pain and itch
  • Dextran sulfate is speculated to have antiviral and antioxidant properties

Application

  • Dextran sulfate is suitable for biochemistry and cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name Dextran sulfate sodium salt
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White to Yellow
Physical Form Powder
Merck Index 142951
Solubility Information Soluble in water.
Quantity 25 g
Formula Weight ≈40 kd
Odor Phenol-like
Chemical Name or Material Dextran sulfate sodium salt
RTECSNumber HH9290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.