Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Dextrin
SCI_ED

Catalog No. s25683 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Solid

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Linear Formula (C6H10O5)n
Quantity 500 g
Formula Weight 162.07
Packaging Solid
Chemical Name or Material Dextrin
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.