Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextromethorphan hydrobromide

Catalog No. p-7042217
Click to view available options
Quantity:
5 g
10 g
50 g

CAS: 125-69-9 | C18H26BrNO | 352.316 g/mol

It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

Solubility
Soluble in water (40 mM), and alcohol (1:10).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-69-9
Molecular Formula C18H26BrNO
Molecular Weight (g/mol) 352.316
MDL Number MFCD02173901
InChI Key MISZALMBODQYFT-URVXVIKDSA-N
Synonym dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
PubChem CID 5464025
SMILES CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br

Specifications

Melting Point 125°C to 127°C
Color White
Quantity 5 g
Beilstein 6453793
Merck Index 14,8091
Solubility Information Soluble in water (40 mM),and alcohol (1:10).
Formula Weight 352.32
Physical Form Solid
Chemical Name or Material Dextromethorphan hydrobromide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-750-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.