Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Dextrose, Monohydrate
SCI_ED

Catalog No. s25296 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 14431-43-7
Synonym Glucose

Specifications

Density ∽1.54g/mL
Packaging Lab Grade
Linear Formula C6H12O6·H2O
Quantity 500 g
Formula Weight 198.18
Grade Laboratory
Chemical Name or Material Dextrose, Monohydrate
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.