Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DFF45/ICAD Antibody [DyLight 650], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4679W
Description
DFF45/ICAD Polyclonal antibody specifically detects DFF45/ICAD in Human samples. It is validated for Western BlotSpecifications
DFF45/ICAD | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
DFF1DNA fragmentation factor 45 kDa subunit, DFF45DNA fragmentation factor subunit alpha, DFF-45DNA fragmentation factor, 45 kD, alpha polypeptide, DNA fragmentation factor, 45kDa, alpha polypeptide, ICAD, Inhibitor of CAD | |
E. coli-derived recombinant human DFF45, Met1-Thr331, Accession # O00273 | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Tumor Suppressors | |
1676 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction