Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536949 Supplier Thermo Scientific Supplier No. PA536949
Add to Cart
Add to Cart

DFNA5 Polyclonal Antibody

DFNA5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DFNA5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Gsdme
Gene Accession No. O60443, Q9Z2D3
Gene Alias 2310037D07Rik; 4932441K13Rik; deafness, autosomal dominant 5; deafness, autosomal dominant 5 (human); deafness, autosomal dominant 5 homolog; Dfna5; DFNA5, deafness associated tumor suppressor; Dfna5h; EG14210; Fin15; Gasdermin-E; Gasdermin-E, C-terminal; Gasdermin-E, N-terminal; GSDME; GSDME-CT; GSDME-NT; ICERE1; ICERE-1; Inversely correlated with estrogen receptor expression 1; nonsyndromic hearing impairment protein; non-syndromic hearing impairment protein 5; Non-syndromic hearing impairment protein 5 homolog
Gene Symbols Gsdme
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the 264-309 of human DFNA5.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1687, 353316, 54722
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.