Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DFNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582429 Supplier Invitrogen™ Supplier No. PA582429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLFDDELLMV LEPVCDDLVS GLSPTVAVLG ELKPRQQQDL VAFLQLVGCS LQGGCPGPED AGSKQLFMTA YFLVSALAEM PDSAAALLGT CCKLQIIPTL CHLLRALSDD GVSDLEDPTL TPLKDTERFG IVQRLFASAD ISLERLKSSV Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 66%.

DFNA5 (deafness, autosomal dominant 5), also known as ICERE-1, is a 496 amino acid protein that is expressed in cochlea tissue, as well as in placenta, brain, heart, liver, lung and pancreas as two alternatively spliced isoforms, designated short and long. Defects in the gene encoding DFNA5 are the cause of non-syndromic sensorineural deafness autosomal dominant type 5 (DFNA5), a form of sensorineural hearing loss that results from damage to one of various structures that receive sound information in the brain. The gene encoding DFNA5 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DFNA5
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gsdme
Gene Accession No. O60443
Gene Alias 2310037D07Rik; 4932441K13Rik; deafness, autosomal dominant 5; deafness, autosomal dominant 5 (human); deafness, autosomal dominant 5 homolog; Dfna5; DFNA5, deafness associated tumor suppressor; Dfna5h; EG14210; Fin15; Gasdermin-E; Gasdermin-E, C-terminal; Gasdermin-E, N-terminal; GSDME; GSDME-CT; GSDME-NT; ICERE1; ICERE-1; Inversely correlated with estrogen receptor expression 1; nonsyndromic hearing impairment protein; non-syndromic hearing impairment protein 5; Non-syndromic hearing impairment protein 5 homolog
Gene Symbols Gsdme
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DFNA5. Recombinant protein control fragment (Product #RP-102091).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1687
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.