Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DFNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103976 Supplier Invitrogen™ Supplier No. PA5103976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DFNA5/GSDME.

DFNA5 (deafness, autosomal dominant 5), also known as ICERE-1, is a 496 amino acid protein that is expressed in cochlea tissue, as well as in placenta, brain, heart, liver, lung and pancreas as two alternatively spliced isoforms, designated short and long. Defects in the gene encoding DFNA5 are the cause of non-syndromic sensorineural deafness autosomal dominant type 5 (DFNA5), a form of sensorineural hearing loss that results from damage to one of various structures that receive sound information in the brain. The gene encoding DFNA5 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DFNA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gsdme
Gene Accession No. O60443, Q9Z2D3
Gene Alias 2310037D07Rik; 4932441K13Rik; deafness, autosomal dominant 5; deafness, autosomal dominant 5 (human); deafness, autosomal dominant 5 homolog; Dfna5; DFNA5, deafness associated tumor suppressor; Dfna5h; EG14210; Fin15; Gasdermin-E; Gasdermin-E, C-terminal; Gasdermin-E, N-terminal; GSDME; GSDME-CT; GSDME-NT; ICERE1; ICERE-1; Inversely correlated with estrogen receptor expression 1; nonsyndromic hearing impairment protein; non-syndromic hearing impairment protein 5; Non-syndromic hearing impairment protein 5 homolog
Gene Symbols Gsdme
Host Species Rabbit
Immunogen A synthesized peptide derived from human DFNA5(Accession O60443), corresponding to amino acid residues L403-A453.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1687, 54722
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.