Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DFNB31 Monoclonal Antibody (2D12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524547
Description
Peptide Sequence: GLLEPTSTLV RVKKSAATLG IAIEGGANTR QPLPRIVTIQ RGGSAHNCGQ LKVGHVILEV NGLTLRGKEH REAARIIAEA FKTKDRDYID FLVTEFNVML.
This gene is thought to function in the organization and stabilization of sterocilia elongation and actin cystoskeletal assembly, based on studies of the related mouse gene. Mutations in this gene have been associated with autosomal recessive non-syndromic deafness and Usher Syndrome. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
DFNB31 | |
Monoclonal | |
Unconjugated | |
WHRN | |
1110035G07Rik; autosomal recessive deafness type 31 protein; AW122018; AW742671; C430046P22Rik; CASK-interacting protein CIP98; CIP98; deafness, autosomal recessive 31; Dfnb31; KIAA1526; PDZD7B; USH2D; whirler; whirlin; Whrn; wi | |
Mouse | |
Protein A | |
RUO | |
25861 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry | |
2D12 | |
PBS with no preservative; pH 7.4 | |
Q9P202 | |
WHRN | |
DFNB31 (NP_056219, 808 a.a. ~ 907 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction