Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DFNB31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116523 Supplier Invitrogen™ Supplier No. PA5116523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, HeLa, K-562, Mouse liver Immunogen sequence: PSGHPDQTGT NQHFVMVEVH RPDSEPDVNE VRALPQTRTA STLSQLSDSG QTLSEDSGVD AGEAEASAPG RGRQSVSTKS RSSKELPRNE RPTDGANKPP GLLEPTSTLV RVKKSAATLG IAIEGGANTR QPLPRIVTIQ RGGSAHNCGQ LKVGHVILEV NGLTLRGKEH REAARIIAEA FKTKDRDYID FLVTEFNVML.

This gene is thought to function in the organization and stabilization of sterocilia elongation and actin cystoskeletal assembly, based on studies of the related mouse gene. Mutations in this gene have been associated with autosomal recessive non-syndromic deafness and Usher Syndrome. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DFNB31
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene WHRN
Gene Accession No. Q80VW5, Q9P202
Gene Alias 1110035G07Rik; autosomal recessive deafness type 31 protein; AW122018; AW742671; C430046P22Rik; CASK-interacting protein CIP98; CIP98; deafness, autosomal recessive 31; Dfnb31; KIAA1526; PDZD7B; USH2D; whirler; whirlin; Whrn; wi
Gene Symbols WHRN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 708-907 of human WHRN (NP_0562193).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25861, 73750
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.