Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DFNB31, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551714
Description
Necessary for elongation and maintenance of inner and outer hair cell stereocilia in the organ of Corti in the inner ear.
Immunogen sequence: VDPGSEAEGS GLKVGDQILE VNGRSFLNIL HDEAVRLLKS SRHLILTVKD VGRLPHARTT VDETKWIASS RIRETMANSA GFLGDLTTEG INKPGFYKGP AGSQVTLSSL GNQTRVLLEE QARHLLNEQE HHighest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifications
| DFNB31 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P202 | |
| WHRN | |
| Recombinant protein corresponding to Human DFNB31 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| WHRN | |
| Whirlin, Autosomal recessive deafness type 31 protein, DFNB31, KIAA1526, WHRN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25861 | |
| Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction