Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGAT1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11041487MFV500
Description
DGAT1 Polyclonal antibody specifically detects DGAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoblotSpecifications
DGAT1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoblotting | |
ACAT-related gene product 1, AGRP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20 | |
A synthetic peptide made to an internal region (within residues 200-300) of human DGAT1. [Swiss-Prot# O75907] | |
0.1 mL | |
Cancer, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
8694 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Immunoblot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction