Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGAT1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10057086
Description
DGAT1 Polyclonal specifically detects DGAT1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
DGAT1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ACAT-related gene product 1, AGRP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20 | |
Goat | |
55.3 kDa | |
0.1 mg | |
Lipid and Metabolism, Lipid Droplets | |
8694 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.1 ug/mL, Flow Cytometry 10 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:64000 | |
O75907 | |
DGAT1 | |
Peptide with sequence C-QNSMKPFKDMDYS corresponding to internal region according to NP_036211.1. | |
Affinity Purified | |
RUO | |
Primary | |
Predicted cross-reactivity based on sequence identity: Mouse and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction