Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGAT1/Diglyceride acyltransferase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984640
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4640 Supplier Bioss Supplier No. BS2332R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.

Specifications

Antigen DGAT1/Diglyceride acyltransferase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DGAT1
Gene Accession No. O75907
Gene Alias ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; DGAT; DGAT1; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; Diacylglycerol O-acyltransferase 1; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; retinol O-fatty-acyltransferase
Gene Symbols DGAT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DGAT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8694
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.