Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DGAT1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518634
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.
Specifications
DGAT1 | |
Polyclonal | |
Unconjugated | |
DGAT1 | |
ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; Dgat; DGAT1; dgat1a; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase 1a; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; hypothetical protein LOC325875; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase; wu:fd36g11; zgc:77691 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
13350, 8694 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75907, Q9Z2A7 | |
DGAT1 | |
Synthetic peptide sequence (QNSMKPFKDMDYS) corresponding to the internal amino acids of DGAT1 (aa 305-317). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction