Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115115 Supplier Thermo Scientific Supplier No. PA5115115
Add to Cart
Add to Cart

DGAT1 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DGAT1
Gene Accession No. O75907, Q9ERM3, Q9Z2A7
Gene Alias ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; Dgat; DGAT1; dgat1a; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase 1a; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; hypothetical protein LOC325875; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase; wu:fd36g11; zgc:77691
Gene Symbols DGAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DGAT1(Accession O75907), corresponding to amino acid residues L105-A155.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13350, 84497, 8694
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.