Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGAT2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171701AF750
Description
DGAT2 Polyclonal specifically detects DGAT2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DGAT2 | |
Polyclonal | |
Western Blot | |
diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL | |
Rabbit | |
0.1 mL | |
84649 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
DGAT2 | |
A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7] | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction