Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DGAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575616
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Acyl-coenzyme A:diacylglycerol acyltransferase, also known as DGAT1 and ARGP1, is a microsomal enzyme that assists in the synthesis of fatty acids into triglycerides. DGAT1 catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol (DAG) and fatty acyl CoA as substrates. DGAT1 plays a fundamental role in the metabolism of cellular diacylglycerol and is important in higher eukaryotes for physiologic processes involving triacylglycerol metabolism, such as intestinal fat absorption, lipoprotein assembly, adipose tissue form-ation and lactation. DGAT2, which has no homology to DGAT1, differs from DGAT1 in that its activity has been shown to be inhibited by MgCl in an in vitro assay. DGAT2 is expressed primarily in liver and white adipose tissue, which suggests that it plays an important role in mammalian triglyceride metabolism.
Specifications
DGAT2 | |
Polyclonal | |
Unconjugated | |
DGAT2 | |
0610010B06Rik; Acyl-CoA retinol O-fatty-acyltransferase; ARAT; DGAT2; DGAT-2; diacylglycerol acyltransferase 2; diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; diglyceride acyltransferase 2; DKFZp686A15125; GS1999FULL; HMFN1045; LOW QUALITY PROTEIN: diacylglycerol O-acyltransferase 2; putative diacylglycerol O-acyltransferase; retinol O-fatty-acyltransferase; UNQ738/PRO1433 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
252900, 67800, 84649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5FVP8, Q96PD7, Q9DCV3 | |
DGAT2 | |
Synthetic peptide corresponding to residues in Human DGAT2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction