Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR14 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP236530AF647
Description
DGCR14 Polyclonal antibody specifically detects DGCR14 in Human, Mouse samples. It is validated for Western BlotSpecifications
DGCR14 | |
Polyclonal | |
Western Blot | |
DGCR13, DGS-HDGS-I, DGSIDGSH, DiGeorge syndrome critical region 13, DiGeorge syndrome critical region 14, DiGeorge syndrome critical region gene 13, DiGeorge syndrome critical region gene 14, DiGeorge syndrome gene H, DiGeorge syndrome gene I, DiGeorge syndrome protein H, ES2DiGeorge syndrome critical region gene DGSI, Es2el, Protein DGCR13, protein DGCR14, Protein ES2 | |
The immunogen recognized by the antibody maps to a region between residue 426 to 476 of human DiGeorge Syndrome Critical Region Gene 14 using the numbering given in entry NP_073210.1. | |
0.1 mL | |
Neuroscience | |
8220 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction