Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551542 Supplier Invitrogen™ Supplier No. PA551542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASASSLLLPA ASRPPRKREA GEAGAATSKQ RVLDEEEYIE GLQTVIQRDF FPDVEKLQAQ KEYLEAEENG DLERMRQIAI KFGSALGKMS REPPPPYVTP ATFETPEVHA GTGVVGNKPR PRGRGLEDGE AGEEEEKEPL PSLDV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

May be involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESS2
Gene Accession No. Q96DF8
Gene Alias AI462402; D16H22S1269E; Dgcr1; DGCR13; DGCR14; DGSH; DGS-H; DGSI; DGS-I; DiGeorge syndrome critical region 13; DiGeorge syndrome critical region 14; diGeorge syndrome critical region 14 homolog; DiGeorge syndrome critical region gene 13; DiGeorge syndrome critical region gene 14; DiGeorge syndrome critical region gene 14 homolog; DiGeorge syndrome critical region gene DGSI; DiGeorge syndrome critical region protein 14; DiGeorge syndrome gene H; DiGeorge syndrome gene I; diGeorge syndrome protein H; ES2; ES2 protein; Es2el; ESS2; Expressed sequence 2 embryonic lethal; Protein DGCR13; protein DGCR14; Protein ES2; Splicing factor ESS-2 homolog
Gene Symbols ESS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DGCR14. Recombinant protein control fragment (Product #RP-91370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.