Learn More
Invitrogen™ DGCR14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589050
Description
Immunogen sequence: METPGASASS LLLPAASRPP RKREAGEAGA ATSKQRVLDE EEYIEGLQTV IQRDFFPDVE KLQAQKEYLE AEENGDLERM RQIAIKFGSA LGKMSREPPP PYVTPATFET PEVHAGTGVV GNKPRPRGRG LEDGEAGEEE EKEPLPSLDV FLSRYTSEDN ASFQEIMEVA KERSRARHAW LYQAEEEFEK RQKDNLELPS AEHQAIESSQ ASVETWKYKA KNSLMYYPEG VPDEEQLFKK PRQVVHKNTR FLRDPFSQAL SRCQLQQAAA LNAQHKQGKV GPDGKELIPQ ESPRVGGFGF; Positive Samples: A-549, NIH/3T3, Mouse testis, Mouse thymus; Cellular Location: Nucleus.
May be involved in pre-mRNA splicing.
Specifications
DGCR14 | |
Polyclonal | |
Unconjugated | |
ESS2 | |
AI462402; D16H22S1269E; Dgcr1; DGCR13; DGCR14; DGSH; DGS-H; DGSI; DGS-I; DiGeorge syndrome critical region 13; DiGeorge syndrome critical region 14; diGeorge syndrome critical region 14 homolog; DiGeorge syndrome critical region gene 13; DiGeorge syndrome critical region gene 14; DiGeorge syndrome critical region gene 14 homolog; DiGeorge syndrome critical region gene DGSI; DiGeorge syndrome critical region protein 14; DiGeorge syndrome gene H; DiGeorge syndrome gene I; diGeorge syndrome protein H; ES2; ES2 protein; Es2el; ESS2; Expressed sequence 2 embryonic lethal; Protein DGCR13; protein DGCR14; Protein ES2; Splicing factor ESS-2 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27886, 8220 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.99 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O70279, Q96DF8 | |
DGCR14, ESS2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DGCR14 (NP_0732101). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.