Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113825 Supplier Invitrogen™ Supplier No. PA5113825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AESCYEKSSF LCKRSQTCVD IKDNVVDEGF YFTPKGDDPC LSCTCHGGEP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DGCR2
Gene Accession No. P98153
Gene Alias 9930034O06Rik; Dgcr2; Dgsc; DGS-C; DiGeorge syndrome critical region gene 2; DiGeorge syndrome critical region protein 2; DiGeorge syndrome gene c; DiGeorge syndrome protein C; IDD; integral membrane protein deleted in DiGeorge syndrome; integral membrane protein DGCR2/IDD; KIAA0163; Lan; mKIAA0163; seizure-related membrane-bound adhesion protein; Sez12; Sez-12
Gene Symbols DGCR2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DGCR2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360742, 9993
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.