Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR6L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113426
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113426 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113426 Supplier Invitrogen™ Supplier No. PA5113426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DGCR6L Polyclonal Antibody for Western Blot, ELISA

This gene, the result of a duplication at this locus, is one of two functional genes encoding nearly identical proteins that have similar expression patterns. The product of this gene is a protein that shares homology with the Drosophila gonadal protein, expressed in gonadal tissues and germ cells, and with the human laminin gamma-1 chain that functions in cell attachment and migration. This gene is located in a region of chromosome 22 implicated in the DiGeorge syndrome, one facet of a broader collection of anomalies referred to as the CATCH 22 syndrome. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR6L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DGCR6L
Gene Accession No. Q9BY27
Gene Alias DGCR6; DGCR6L; DiGeorge syndrome critical region 6-like protein; DiGeorge syndrome critical region gene 6 like; DiGeorge syndrome critical region gene 6-like; protein DGCR6L
Gene Symbols DGCR6L
Host Species Rabbit
Immunogen Recombinant Human Protein DGCR6L protein (1-220AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85359
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.