Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR8 Antibody (2H2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DGCR8 |
---|---|
Clone | 2H2 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
DGCR8 Monoclonal antibody specifically detects DGCR8 in Human samples. It is validated for Western Blot, ELISASpecifications
DGCR8 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha | |
A synthesized peptide derived from Human DGCR8 [UniProt Q8WYQ5] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2H2 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Epigenetics, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
54487 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title