Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR8 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130116V
Description
DGCR8 Polyclonal antibody specifically detects DGCR8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DGCR8 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha | |
Antigen maps to a region between residue 400 and 450 of human DiGeorge syndrome critical region gene 8 using the numbering given in entry NP_073557.3 (GeneID 54487). | |
0.1 mL | |
Epigenetics, Stem Cell Markers | |
54487 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction