Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | DGCR8 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DGCR8 Polyclonal antibody specifically detects DGCR8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DGCR8 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Epigenetics, Stem Cell Markers | |
PBS, pH 7.2, 40% glycerol | |
54487 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha | |
This antibody was developed against Recombinant Protein corresponding to amino acids: YGASLLSKGSFSKGRLLIDPNCSGHSPRTARHAPAVRKFSPDLKLLKDVKISVSFTESCRSKDRKVLYTGAERDVRAECGLLLSPVSGDVHACPFGGSVGDGVGIGGESADKKDEENELDQEKRVEYAVLDELEDFTDNLELDEEGAGGFTAKAIVQRDRVDEEALNFPYEDDFDNDVDAL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title