Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578510 Supplier Invitrogen™ Supplier No. PA578510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431 Predicted Reactivity: Mouse (95%), Rat (92%), Pig (93%), Rhesus Monkey (99%), Bovine (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene DGCR8
Gene Accession No. Q8WYQ5, Q9EQM6
Gene Alias C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06
Gene Symbols DGCR8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human DGCR8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54487, 94223
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.