Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR8 Recombinant Rabbit Monoclonal Antibody (3T5Q6)
GREENER_CHOICE

Catalog No. PIMA538100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538100 Supplier Invitrogen™ Supplier No. MA538100
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, RD, U-251MG, Mouse brain Immunogen sequence: DLEEFRSYLE KRFDFEQVTV KKFRTWAERR QFNREMKRKQ AESERPILPA NQKLITLSVQ DAPTKKEFVI NPNGKSEVCI LHEYMQRVLK VRPVYNFFEC ENPSEPFGAS VTIDGVTYGS GTASSKKLAK NKAARATLEI LIPDFVKQTS EEKPKDSEEL EYFNHISIED SRVYELTSKA GLLSPYQILH ECLKRNHGMG DTSIKFEVVP GKNQKSEYVM ACGKHTVRGW CKNKRVGKQL ASQKILQLLH PHVKNWGSLL RMYGRESSKM VKQETSDKSV IELQQYAKKN KPNLHILSKL QEEMKRLAEE REETRKKPKM SIVASAQPG.

This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR8
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3T5Q6
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DGCR8
Gene Accession No. Q8WYQ5, Q9EQM6
Gene Alias C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06
Gene Symbols DGCR8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 436-764 of human DGCR8 (Q8WYQ5).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54487, 94223
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.