Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGCR8 Recombinant Rabbit Monoclonal Antibody (ST04-79)
GREENER_CHOICE

Catalog No. PIMA532153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532153 Supplier Invitrogen™ Supplier No. MA532153
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR8
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone ST04-79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene DGCR8
Gene Accession No. Q8WYQ5, Q9EQM6
Gene Alias C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06
Gene Symbols DGCR8
Host Species Rabbit
Immunogen Recombinant protein within Human DGCR8 aa 383-773.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287954, 54487, 94223
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.