Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGK-alpha Antibody (OTI4A11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270547B
Description
DGK-alpha Monoclonal specifically detects DGK-alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
DGK-alpha | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
DGKA | |
Full length recombinant protein of human DGK-alpha (NP_001336) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4A11 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
1606.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction